missing translation for 'onlineSavingsMsg'
Learn More

Sorbitan monolaurate, Thermo Scientific Chemicals

£26.13 - £65.12

Chemical Identifiers

CAS 1338-39-2
Molecular Formula C18H34O6
Molecular Weight (g/mol) 346.46
MDL Number MFCD00005365
InChI Key LWZFANDGMFTDAV-IOVMHBDKSA-N
Synonym Sorbitan monododecanoate; Span™ 20
IUPAC Name 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11338615
SDS View Documents
Thermo Scientific Alfa Aesar
L12099.30
250 g
£26.13
250g
Please to purchase this item. Need a web account? Register with us today!
11348615
SDS View Documents
Thermo Scientific Alfa Aesar
L12099.0B
1000 g
£65.12
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sorbitan monolaurate is a detergent and monoester of lauric acid and hexitol anhydrides derived from sorbitol. It is used in a study to assess enhanced solubility and oral bioavailability of itraconazole. It is also used in a study to investigate the oxidation of unsaturated lipids in oil-in-water emulsions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sorbitan monolaurate is a detergent and monoester of lauric acid and hexitol anhydrides derived from sorbitol. It is used in a study to assess enhanced solubility and oral bioavailability of itraconazole. It is also used in a study to investigate the oxidation of unsaturated lipids in oil-in-water emulsions.

Solubility
Soluble in 2-ethoxyethanol, ethanol and methanol. Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-663-3

missing translation for 'rtecsNumber' : WG2920000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sorbitan monolaurate, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.