missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sorbitol MacConkey Agar

Product Code. 12922608
Click to view available options
Description:
Sorbitol MacConeky Agar
This item is not returnable. View return policy

Product Code. 12922608

Brand: Thermo Scientific™ PO0232A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selectively detect Escherichia coli O157 with this selective, differential and ready-to-use medium.

Selectively detect Escherichia coli O157 with Thermo Scientific™ Oxoid™ Sorbitol MacConkey Agar. The formulation, based on that described by Rappaport and Henig1 is identical to MacConkey Agar No.3, except that lactose has been replaced with sorbitol. E. coli O157 does not ferment sorbitol and, therefore, produces colorless colonies. In contrast, most E. coli strains ferment sorbitol and form pink colonies.

Escherichia coli O157 is a significant human pathogen and is recognised as a cause of hemorrhagic colitis, an illness characterized by bloody diarrhoea, severe abdominal pain and hemolytic uraemic syndrome. Use Sorbitol MacConkey Agar for selective detection of Escherichia coli O157.

  • Sensitive and specific for E. coli O157 based on sorbitol fermentation.
  • Recommended as a simple, inexpensive, rapid and reliable means of screening for E. coli O157.
  • Ready-to-use convenience of prepared media.

The efficiency of Sorbitol MacConkey Agar has been confirmed by March and Ratnam2. These workers reported a sensitivity of 100% and a specificity of 85%, and recommended the medium as a simple, inexpensive, rapid and reliable means of screening for E. coli O157.

The selectivity of the medium is further enhanced by the addition of cefixime and potassium tellurite. The level of potassium tellurite selects serogroup O157 from other E. coli serogroups and inhibits Providencia and Aeromonas species. Cefixime is inhibitory to Proteus spp.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Dark Red
Content And Storage 2°C to 10°C
Description Sorbitol MacConeky Agar
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 19.5g±1.0.g
Packaging Type 10 x 90mm plates, film wrapped
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.