Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Buckets for 4-Place Swinging Bucket Rotors
Round or rectangular buckets for use with Four-Place Swinging Bucket Rotors 75006445 and 75006434
Brand:  Thermo Scientific™ 75006446
 View more versions of this product
Product Code. 10638952
  • £477.00 / Each
Explore more special offers
Description
Description
Order Info
Sold individually; order correct quantity to equip rotor. Sealing lids sold separately. Accessory adapters accept assorted tube styles and volumes from 1.5 to 250mL.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Swinging bucket rotor
Rectangular Bucket, 1 x 250 mL