Learn More
3M™ Sound Examiner 400 Series Sound Level Meter SE 401
3M™ Sound Examiner 400 Series Sound Level Meter SE 401 is engineered to accurately measure noise levels in highly variable environments.
Brand:  3M™ 7100010228
Product Code. 15730028
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Sound Level Meter engineered to accurately measure noise levels in highly variable environments
  • Intuitive interface allows for out of the box use with little or no training
  • Preamp can be easily removed to enable interference-free low level monitoring
  • Class 1
  • Rechargeable lithium battery
  • Store numerous noise surveys in memory for download
  • Multilingual user interface
TRUSTED_SUSTAINABILITY
Specifications
Specifications
18 hours
28.4 x 7.1 x 3.3 cm
Sound Level Meter
Area Monitoring (Sound Level Meter)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Sound Examiner 400 Series Sound Level Meter SE 401 > Battery Life: 18 hours