missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Soundproof Box for Ultrasonic Baths
Reduces the typical cavitation noise of an operating ultrasonic unit to a tolerable level. Fisherbrand™ Soundproof Box for Ultrasonic Water Baths allows operation of ultrasonic cleaning units in sensitive work areas such as chemical and dental laboratories, medical practices or even workshops.
Brand: Fisherbrand™ 2000001567
This item is not returnable.
View return policy
Description
- Noise reduction by approx. 20dBA
- Feels like a noise reduction of 75%
- Vapours carried off by an exhaust device to protect the box inside against accumulation of heat or humidity
Specifications
Nap Mat | |
Silicone |