Learn More
Fisherbrand™ Sound Protection Cover for Elmasonic X-tra ST Ultrasonic Cleaners
Reduces the noise generated by ultrasonic cleaning units while providing a secure closure. For use with Ultrasonic Water Baths.
Brand: Fisherbrand™
Code : NEW
Additional Details : Weight : 1.00000kg
Description
- Hinged design
- Available in a range of sizes for all X-tra ST models
- Manufactured from stainless steel
- Accessory item
Specifications
Sound Protection Cover | |
Hinged Lid | |
Stainless Steel |
Ultrasonic Cleaner, Elmastonic Xtra ST 300 H/500 H | |
Noise reduction for ultrasonic cleaning units |