missing translation for 'onlineSavingsMsg'
Learn More

Cytiva SOURCE™ 30S Chromatography Media

Product Code. 11500814 Shop All Cytiva Products
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
This item is not returnable. View return policy

Product Code. 11500814

Brand: Cytiva 17127301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemically stable cation exchanger for high resolution polishing purification of proteins, peptides and oligonucleotides

  • Average particle size: 30μm
  • Ligand: sulphonate group
  • Matrix: polystyrene/divinylbenzene
  • pH stability working range: 2 to 13
  • Available in 50mL bottle

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mL
Stability Stable in common Ion Exchange Buffers
For Use With (Application) For High Resolution Polishing PurifiCatIon of Proteins, Peptides and Oligonucleotides
Storage Requirements 4°C to 30°C
Product Type Source 30 S chromatography media
Flow Rate 2000 cm/Hoμr
Particle Size 30 μm
Height (English) Bed 3.93 in.
Height (Metric) Bed 10 cm
Max. Pressure 1000 kPa
Matrix Polystyrene/Divinylbenzene
pH Range 2 to 13
Storage Buffer 20% Ethanol, 0.2 M Sodium Acetate
Ligand Type Sulphonate Group
Show More Show Less
compliance-icons
Product Identifier
  • SOURCE 30S, 50 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.