missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX2 Mouse anti-Human, Mouse, Porcine, Clone: 4B9B10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66411-1-IG-150UL
This item is not returnable.
View return policy
Description
This antibody is raised against an internal region of human SOX2.
SOX2 is an intronless gene encoding a member of the SRY-related HMG-box (SOX) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The product of the SOX2 gene is required for stem-cell maintece in the central nervous system, and also regulates gene expression in the stomach. The SOX2 gene lies within an intron of another gene called SOX2 overlapping transcript (SOX2OT). Further, SOX2 protein may act as a transcriptional activator after forming a protein complex with other proteins. Mutations in the SOX2 gene have been associated with bilateral anophthalmia, a severe form of structural eye malformation, optic nerve hypoplasia and syndromic microphthalmia.Specifications
SOX2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P48431, P48432 | |
SOX2 | |
SOX2 Fusion Protein Ag13635 | |
150 μL | |
Primary | |
Human, Mouse, Porcine | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
4B9B10 | |
Unconjugated | |
SOX2 | |
ANOP3, MCOPS3, SOX2, Transcription factor SOX 2 | |
Mouse | |
Protein G | |
RUO | |
20674, 407739, 6657 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction