missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ SP Quick Single-channel Pump Heads

Fast and convenient tube changes

£573.00 - £647.00

Specifications

Flow Rate 0.38 to 3436 mL/min.
Type Pump Head
View More Specs

Products 2
Product Code Brand For Use With Price Quantity & Availability  
Product Code Brand For Use With Price Quantity & Availability  
10130791
View Documents Promotion Details
Heidolph™
527-11100-00
Single-channel pumps; tubing (wt 1.6 mm)
£647.00
Each
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
 
10439882
View Documents Promotion Details
Heidolph™
527-11300-00
Single-channel pumps; tubing (wt 2.5 mm)
£573.00
Each
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
 
Description

Description

  • Low pulsation due to 5 roller system
  • Pump head features ball bearings
  • Rollers made of stainless steel
  • Depending on drive and tubing, flow rate ranges from 0.38 to 3436 mL/min.
Specifications

Specifications

0.38 to 3436 mL/min.
Pump Head
Videos
SDS
Documents

Documents

Product Certifications