Learn More
Wilmad-LabGlass™ SP Scienceware™ 10mm OD High-Throughput NMR Tubes
Designed for routine use with small molecule (MW<500) samples, Wilmad-LabGlass 10mm OD High-Throughput NMR tubes are ideal for use in most low to mid field NMR spectrometers (up to 600 MHz).
Brand: Wilmad-LabGlass™ 66000-0081
Code : 03
Additional Details : Weight : 0.86170kg
Alert:
Note: For NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, please refer to Wilmad Precision NMR Tubes.
Description
- Manufactured from borosilicate glass that meets ASTM Type 1 Class B standard and recognized as N51A
- This glass may contain Fe2O3 which is paramagnetic and can impact shimming quality at high field
- Guaranteed spectral resolution for small molecule (MW<500) samples up to 600 MHz
- Averaged camber of 60 micron
- Ideal for MRI imaging applications
- Blue disposable NMR caps included; other colors available separately
- 100% inspected for surface defects and physical dimension to ensure the success of your experiments
Specifications
High-Throughput NMR Tube | |
60 μm | |
17.8 cm | |
9.944±0.025 mm | |
Blue disposable NMR caps | |
0.02 in. | |
100 |
Borosilicate Glass | |
Low to mid field NMR spectrometers (up to 600 MHz) | |
Clear | |
Small molecule (MW<500) samples | |
7 in. | |
0.60 mm | |
WG-4000-7 |