Learn More
Bel-Art™ SP Scienceware™ LabMat™ Bench Liners
Simplify cleanup and safeguard work surfaces from stains, spills and wear. Bel Art™ SP Scienceware™ LabMat™ Bench Liners can be rolled up and discard after contamination.
Brand: Bel-Art™ F24675-1000
106.56 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Code : 02
Additional Details : Weight : 0.86850kg
Description
- Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
- Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
- Cut to size easily with scissors or the Scienceware™ Safety Cutter (Mfr. No. 21012-0000)
- Roll up and discard in proper waste container after contamination
- Select orange to identify use with biohazard materials
Specifications
Bench Protector | |
400 mL/m2 | |
15.2 m | |
508 mm | |
Orange | |
Yes | |
Each |
Polyethylene | |
50 ft. | |
20 in. | |
50 ft. x 20 in. (15.2m x 508mm) | |
One single roll, 50 ft. | |
Simplify cleanup and safeguard work surfaces from stains, spills and wear |