missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Reusable Polypropylene Scoops

Designed for retrieving samples from a larger source. SP Scienceware™ Reusable Polypropylene Scoops are a valuable multi-purpose lab tool.

Brand:  Bel-Art™ F36750-0000

 View more versions of this product

Product Code. 11746545


Description

Description

Designed for retrieving samples from a larger source. SP Scienceware™ Reusable Polypropylene Scoops are a valuable multi-purpose lab tool.

  • Rigid form scoop
  • Flat bottom keeps scoops upright when filled or empty
  • Can be used as weighing boat 
  • Manufactured from polypropylene
  • Steam autoclavable at 121°C (250°F)
  • Complies with FDA regulations for food contact
  • Capacity/volumes are estimated and are not meant for taking exact sample weights/volumes
  • Stated volumes represent typical heaping samples
Specifications

Specifications

Scoop
Autoclavable
125 mL
One-piece
Smooth
Designed to remain upright when filled or empty allowing them to be used as weighing boats or to be placed on a flat surface for ease sample collection and dispensing
1.31 in.
8.69 in.
22.1 cm
2 in.
Polypropylene
4 oz.
Reusable
Translucent
FDA
22.1 x 5.1 x 3.3 cm
3.3 cm
5 in.
12.7 cm
5.1 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers