missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel Art™ Secador™ Standard Desiccator Cabinet, Horizontal Model 1.0
Constructed of Durastar™ co-polyester. Standard desiccator cabinets block 99% of UV light and are resistant to staining, crazing, and chemical attack.
£320.00 - £388.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Cabinet Style | Vertical |
Material | Copolyester |
Product Type | Desiccator |
Door Style | Left-hinged and gasketed |
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
10760583
|
Bel Art™
F42071-0006 |
Clear with Blue Ends | N/A | N/A | N/A | |||||
10086471
|
Bel Art™
F42071-0000 |
Clear |
£388.00
Each |
Estimated Shipment: 03-12-2025
Log in to see stock. |
||||||
Description
- Amber color model reduces visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
- Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
- Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
- Stack up to 3 units high to save space
- Can be used with desiccant cartridges, F420450250, F420450200, F420480100
Specifications
Not Autoclavable | |
Copolyester | |
Left-hinged and gasketed | |
1.0 | |
Chemicals, stains and fine cracks | |
0.75 cu. ft. | |
16.3 in. | |
13.4 in. | |
34.1 cm | |
4.5 in. | |
16.3 in. |
Vertical | |
Desiccator | |
Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250 | |
1 | |
Fixed Perforated | |
16.3 x 13.4 in. (41.4 x 34.1 cm) | |
41.4 cm | |
8.9 in. | |
8.4 in. | |
21.3 cm | |
41.4 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bel Art™ Secador™ Standard Desiccator Cabinet, Horizontal Model 1.0