missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Secador™ Standard Desiccator Cabinets: Horizontal Models
A UV light-blocking, dry environment
£819.00 - £844.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Cabinet Style | Horizontal |
Material | Copolyester |
Door Style | Bottom-hinged and gasketed |
For Use With (Application) | Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250 |
Product Code | Brand | Color | Shelf Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Shelf Type | Price | Quantity & Availability | ||||
11377184
|
Bel-Art™
F42074-0000 |
Clear | Adjustable Perforated |
£819.00
Each |
Estimated Shipment: 21-01-2025
Log in to see stock. |
||||
10178751
|
Bel-Art™
F42074-0006 |
Clear with Blue End Caps | Vented Adjustable |
£844.00
Each |
In Stock
Log in to see stock. |
||||
11802732
|
Bel-Art™
F42074-0007 |
Blue with Clear Door | Vented Adjustable | ||||||
Description
The attractive design and sturdy construction of the Secador line of desiccator cabinets assures easy and reliable storage of moisture and light sensitive products.
- Durastar™ co-polyester construction blocks 99% of UV light and resists staining, crazing and chemical attack
- Amber color models reduce visible light penetration by over 50%, making them outstanding choices for storing light sensitive materials like reagents, analytical standards and investigational compounds
- Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
- Perforated shelves facilitate circulation of dry air; Space for desiccant under bottom shelf
- Easy-to-view hygrometer is located in the door front
- 4.0 horizontal has a wider footprint, lower height and pull down “oven style” door
- U.S. Patents: 6,834,920; 7,318,630 and D474,626
Dimensions
- Secador 1.0: 0.74 cu. ft. internal volume, 16.3L x 13.4W x 8.4 in.H
- Secador 2.0: 1.16 cu. ft. internal volume, 16.3L x 13.4W x 12.4 in.H
- Secador 3.0: 1.55 cu. ft. internal volume, 16.3L x 13.4W x 16.4 in.H
- Secador 4.0: 1.87 cu. ft. internal volume, 16.3L x 13.4W x 20.4 in.H
Order Info
Replacement shelves are available: Replacement Shelf for Horizontal 4.0 Cabinets, Mfr. No. 420740001 or Replacement Shelf for Vertical 1.0, 2.0, 3.0 or 4.0 Cabinets, Mfr. No. 420741001. Use with desiccant cartridges Mfr. Nos. 420450250, 420450200, 420480100.
Specifications
Not Autoclavable | |
Copolyester | |
Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250 | |
4.0 | |
Chemicals, stains and fine cracks | |
8.9 in. | |
16.4 in. | |
16.3 in. |
Horizontal | |
Bottom-hinged and gasketed | |
Carrying handles, 2 latches and loops for a padlock or tamper-evident seals, door-mounted hygrometer | |
2 | |
1.9 cu. ft. | |
13.4 in. | |
34.1 cm | |
41.4 cm |