missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Vibrasorb™ Premium Vibration Damping Mounts
Protects electromechanical and optical equipment from exposure to vibration. Bel Art™ SP Scienceware™ Vibrasorb™ Premium Vibration Damping Mounts facilitates optimal equipment use.
£1155.00 - £1320.96
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12320679
|
Bel-Art™
H18386-0000 |
Premium Vibration Damping Mount; 45 x 56 x 7.6cm |
£1320.96
Each |
In Stock
Log in to see stock. |
|||||
12330679
|
Bel-Art™
H18386-0001 |
Premium Vibration Damping Mount; 35 x 35 x 7.6cm |
£1155.00
Each |
Estimated Shipment: 05-09-2024
Log in to see stock. |
|||||
Description
Protects electromechanical and optical equipment from exposure to vibration. Bel Art™ SP Scienceware™ Vibrasorb™ Premium Vibration Damping Mounts facilitates optimal equipment use.
- Polished surface resists scratches and chemical attack 6.4cm (2½ in.) thick platforms
- Molded of dense black & white terrazzo
- Four Vibro-Absorber™ cushions with neoprene feet support up to 16kg (35 lb)
- Larger size will absorb vibrations down to 13 Hz with a 16kg (35lb) load; smaller size will absorb vibrations down to 16 Hz. With a 16kg (35lb) load
- With lighter loads, the minimum frequency of damping for both sizes will increase to a value between 16 Hz. And 25 Hz
- Shipping weights: large 39kg (87lb), small 19kg (42lb)