missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva SP Sepharose™ High Performance Media
Use for high intermediate and polishing steps in downstream processing. Cytiva Life Sciences™ SP Sepharose™ High Performance Media is composed of crosslinked agarose beads.
Brand: Cytiva 17-1087-01
331.55 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
This item is not returnable.
View return policy
Description
- Composed of crosslinked agarose beads with mean diameter of 34μm, modified with sulphopropyl (SP) strong cation exchange groups
- High resolution, high capacity separation with high recovery
- Chemical stability for effective CIP/sanitization
- Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development
Specifications
75 mL | |
For High Intermediate and Polishing Steps in Downstream Processing | |
SP Sepharose High Performance Media | |
24 to 44 μm | |
10 cm | |
Strong CatIon Exchanger | |
6% Cross-Linked Agarose | |
20% Ethanol, 0.2 M Sodium Acetate |
Stable in commonly used Buffers: 8 M Urea, 6 M Guanidine HCl, 70% Ethanol, 1 M NaOH, 1 M Acetic Acid, 30% Isopropanol, 30% Acetonitrile, 2% SDS1 | |
4°C to 30°C | |
150 cm/Hoμr | |
3.93 in. | |
0.15 to 0.20 mM H+/mL | |
<3 bar | |
3 to 14 | |
Sulphopropyl |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- SP Sepharose High Performance, 75 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ethanol