Learn More
B Braun™ Space PCA Infusion Pump System
Small syringe pump that is big on safety and accuracy in any care setting. B Braun™ Space PCA Infusion Pump System is ideal for operating rooms, adult care, pediatric and emergency settings where ease of use and precise medication administration are critical.
Brand: B Braun™ 8713030
Code : NEW
Additional Details : Weight : 0.99000kg
Description
- Independent Modularity: A microprocessor in each pump helps prevent channel confusion and catastrophic pump failures.
- Easily Stackable: Vertical stacking takes less space for increased versatility.
KeyGuard™: Non-numeric keypad with simple arrows to help prevent manual programming errors related to zero vs. decimal entry. - Intuitive Universal User Interface on all Space Pumps reduces training required by staff.
- Extensive Drug Library with customizable hard and soft limits and clinical advisories
Integrated piston brake helps prevent inadvertent bolus during syringe change. - Post-occlusion bolus reduction software.
- Automatic drive technology ensures accuracy and safety.
- Horizontal and vertical syringe pump mounting options.
- Easy-to-learn, intuitive and user friendly 1,500-entry drug library available.
- Alarms are audible, clearly visible and differentiated using different tones.
- Strong lightweight pole clamp holds up to three pumps mounted horizontally or vertically.
Specifications
Space PCA Infusion Pump System | |
Operating rooms, adult care, pediatric and emergency settings where ease of use and precise medication administration are critical |