Learn More
Tocris Bioscience™ Spadin
Potent K2P2.1 (TREK-1) channel blocker
Brand:  Tocris Bioscience™ 5594/1
Product Code. 18744858
  • £171.00 / 1mg
Explore more special offers
Description
Description
Spadin is a potent TREK-1 channel blocker (IC50 = 71 nM). Enhances dorsal raphe nucleus 5-HT neurotransmission in mice. Induces hippocampal CREB activation and neurogenesis in adult mice. Exhibits antidepressant effects in mouse models of depression. Brain penetrant.
Specifications
Specifications
0.95
2012.34
1 mg
Spadin
Spadin
Tyr-Ala-Pro-Leu-Pro-Arg-Trp-Ser-Gly-Pro-Ile-Gly-Val-Ser-Trp-Gly-Leu-Arg