Learn More
SPANXE Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15870-1-AP-150UL
 View more versions of this product
Product Code. 16825254
  • £349.00 / 150µL
Explore more special offers
Description
Description
Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family of cancer/testis-associated genes, which are located in a cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene encodes a sperm protein that contains a consensus nuclear localization signal but, although a role in spermatogenesis is suggested, the specific function of this family member has not yet been determined.
Specifications
Specifications
SPANXE
Polyclonal
Unconjugated
SPANXD
SPANX A2, SPANX C, SPANX D, SPANX E, SPANX family member E, SPANX family, member E, SPANXA2, SPANXD, SPANXE
Rabbit
Antigen Affinity Chromatography
RUO
64648
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BXN6
SPANXD
SPANXE Fusion Protein Ag8572
150 μL