missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPANXE Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15870-1-AP-20UL
This item is not returnable.
View return policy
Description
Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family of cancer/testis-associated genes, which are located in a cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene encodes a sperm protein that contains a consensus nuclear localization signal but, although a role in spermatogenesis is suggested, the specific function of this family member has not yet been determined.Specifications
SPANXE | |
Polyclonal | |
Unconjugated | |
SPANXD | |
SPANX A2, SPANX C, SPANX D, SPANX E, SPANX family member E, SPANX family, member E, SPANXA2, SPANXD, SPANXE | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64648 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BXN6 | |
SPANXD | |
SPANXE Fusion Protein Ag8572 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |