missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Socorex™ Spare Kit Bag, Socorex™ 187 Syringe
Replace valve assembly parts for the Socorex™ 187 Tube Filling syringe if necessary. Socorex™ Spare Kit Bag, Socorex™ 187 Syringe includes all the spare parts needed for repair/replacement of your Socorex™ 187 Syringe valve.
£6.54 - £8.00
Specifications
Includes | 2/ea. springs, 2/ea. balls, 2/ea. valve washers, one barrel washer. |
---|---|
Product Type | Spare Kit Bag |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10633633
|
Socorex™
1.187.7210 |
Socorex 187 Tube Feeding Syringe, 10 mL |
£8.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10580943
|
Socorex™
1.187.7202 |
Socorex 187 Tube Feeding Syringe, 2 mL |
£7.75
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10631903
|
Socorex™
1.187.72005 |
Socorex 187 Tube Feeding Syringe, 0.5 mL |
£6.54
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Spare parts for Socorex™ 187 Tube Feeding Syringe models based on syringe volume.
- Each bag includes the following:
- 2/ea. springs.
- 2/ea. balls.
- 2/ea. valve washers.
- 1/ea. barrel washer.
Specifications
2/ea. springs, 2/ea. balls, 2/ea. valve washers, one barrel washer. | |
Spare Kit Bag |