missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Spare Burette Tip

Spare Burette Tip for use with compact burettes, automatic burettes, and titrators.

Brand:  BRAND™ 11515

7.05 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


Product Code. 10727444

  • £8.10 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Constructed of amber glass
  • Replacement tip
  • For 25 mL and 50 mL burettes
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Amber
25 and 50 mL Compact and Automatic Burette
1.2 cm
7.1 cm
Borosilicate Glass (5.4)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.