Learn More
Fisherbrand™ Spare Cartridges for Portable Demineralizers
Facilitates the demineralization of water with conductivity of 0.3 to 2μS/cm and pH 6 to 7. Fisherbrand™ Spare Cartridges for Portable Demineralizers start blue and gradually change color to ocher as they become saturated.
Brand:  Fisherbrand™
Code : A0
Additional Details : Weight : 5.85000kg
 View more versions of this product
Product Code. 11964169
  • £282.00 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Contain only new resins that have not been polluted or regenerated
  • Level of performance is constant and guaranteed
  • Robust and practical construction
  • Quick connectors make assembly/dismantling easy and fast
Specifications
Specifications
Blue cartridge turns ocher brown
150 L/hr.
60 cm
5/Set
Depends on the quality of the inlet water
Ion Exchanger
Portable Demineralizer DEM'O5
Spare Cartridge