Learn More
Buerkle™ Spare Latex Hose for Buerkle™ Titrating Burettes
Buerkle™ Spare Latex Hose for Buerkle™ Titrating Burettes.
Brand: Buerkle™ 9695-0041
Additional Details : Weight : 0.01500kg
Description
- Latex
- Spare hose
- For use with Buerkle™ Titrating Burettes
Specifications
Replacement article for titrating burette | |
1000 mm | |
Latex | |
Spare Hose Latex |
Brown | |
Titrating Burette Automatic | |
1000 mm |