Learn More
Lenz Laborglasintrumente Spare Valve Spindles
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 02280002
Product Code. 11566824
  • £19.20 / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Spare spindles for needle-valve stopcocks
  • Material: PTFE (spindle), PP (screw cap and retaining device)
  • Easy to change
  • Pack: 1 piece
Specifications
Specifications
PTFE
Needle-valve stopcocks