missing translation for 'onlineSavingsMsg'
Learn More

Liebherr™ Spark-Free Upright Lab Freezer with Shelves
GREENER_CHOICE

Secure, accurate and ultra energy-efficient, spark-free interior, freezer storage, the Liebherr™ Spark-Free Interior, Lab Freezer is designed for general purpose applications with Liebherr build-quality you can rely on

Brand:  Liebherr™ SFFfg 5501

2632.29 GBP valid until 2024-12-31
Use promo code "21995" to get your promotional price.


 View more versions of this product

Product Code. 17889106

  • £3800.00 / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Spark-Free upright Lab Freezer for storage of solvents and reagents, reach-in, 1-section
  • Temperature range -9 to -30°C, ± 5°C temperature stability tested to EN60068-3
  • White exterior, commercial-grade molded polystyrol interior, static refrigeration with manual defrost
  • Refrigerant R 290, connection rating 2.0 A, 220-240V/50/1-ph
  • This model is intended for the spark-free storage
  • ATEX-certified spark-free interior
  • Multifunctional digital controller with high resolution display and connectivity options
  • Door lock, reversible hinges, antimicrobial lever handle
  • Accessories: optional set of drawers available 
Specifications

Specifications

Atex Freezer
-10°C to -30°C
-20°C
Reversible, Self-closing, Fitted Lock
Scratch-resistant Painted Steel
Commercial-grade Molded Polystyrol
5
Upright
Optional (80 to 100 mm dia.)
Low-maintenance Rear-wall Wire-on-tube
1 x 10 mm dia.
Manual
SmartCoolingHub
CEE 7/7
1, Single Phase
516 kWh/a
R 290
499 L
NTC
±5 K
1
Stainless Steel
White Powder-coated
ISO 9001, ISO 14001, CE
Touch Control, Digital Display in Door
White
Storage of Temperature-sensitive Products
2 yr. Parts and Labor
SFFfg 5501
2.0 A
220/240 V
150 W
CFC-free
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers