missing translation for 'onlineSavingsMsg'
Learn More

Liebherr™ Spark-Free Upright Lab Fridge with Shelves
GREENER_CHOICE

Secure, accurate and ultra energy-efficient, spark-free interior, fridge storage, the Liebherr™ Spark-Free Interior, Lab Fridge is designed for general purpose applications with Liebherr build-quality you can rely on.

Brand:  Liebherr™ SRFFG4001

1820.63 GBP valid until 2025-07-11
Use promo code "25721" to get your promotional price.


 View more versions of this product

Product Code. 17859096

  • £2410.00 / Each
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Spark-Free upright Lab Fridge for storage of solvents and reagents, reach-in, 1-section
  • Temperature range +3 to +16°C, +/- 3°C temperature stability tested to EN60068-3
  • white exterior, commercial-grade molded polystyrol interior, fan-assisted refrigeration
  • Refrigerant R 600a, connection rating 2.0 A, 220-240V/50/1-ph
  • This model is intended for the spark-free storage
  • ATEX-certified spark-free interior
  • Multifunctional digital controller with high resolution display and connectivity options
  • Door lock, reversible hinges, antimicrobial lever handle
Specifications

Specifications

Atex Fridge
3°C to 16°C
+5°C
R 600a
1
Scratch-resistant Painted Steel
Commercial-grade Molded Polystyrol
Upright
Optional (80 to 100 mm dia.)
Low-maintenance Rear-wall Wire-on-tube
1 x 10 mm dia.
2 yr. Parts and Labor
SRFfg 4001
Yes
2.0 A
220/240 V
421 kWh/a
394 L
NTC
±3 K
Solid Reversible Self-closing with Lock
5
White Powder-coated
ISO 9001, ISO 14001, CE
Touch Control, Digital Display in Door
White
Storage of Temperature-sensitive Products
Scientific Performance
Timed Off-cycle
SmartCoolingHub
CFC-free
CEE 7/7
120 W
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.