Learn More
SPDP, MedChemExpress
MedChemExpress SPDP (SPDP Crosslinker) is a short-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls. It is a glutathione cleavable ADC linker used for the antibody-drug conjugates (ADCs).
£75.02 - £1103.52
Specifications
Chemical Name or Material | SPDP |
---|---|
CAS | 68181-17-9 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C12H12N2O4S2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16494256
|
Medchem Express
HY-100216 |
100 mg |
£317.02
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16472848
|
Medchem Express
HY-100216 |
10 mg |
£75.02
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16471516
|
Medchem Express
HY-100216 |
500 mg |
£1103.52
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16440906
|
Medchem Express
HY-100216 |
200 mg |
£543.29
200mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16439506
|
Medchem Express
HY-100216 |
50 mg |
£180.29
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPDP (SPDP Crosslinker) is a short-chain crosslinker for amine-to-sulfhydryl conjugation via NHS-ester and pyridyldithiol reactive groups that form cleavable (reducible) disulfide bonds with cysteine sulfhydryls. It is a glutathione cleavable ADC linker used for the antibody-drug conjugates (ADCs).Specifications
SPDP | |
White | |
C12H12N2O4S2 | |
DMSO : 155 mg/mL (496.22 mM; Need ultrasonic) | |
312.36 | |
97.0% | |
Solid |
68181-17-9 | |
Research | |
SPDP Crosslinker | |
O=C(ON1C(CCC1=O)=O)CCSSC2=NC=CC=C2 | |
312.36 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : -20°C, protect from light, stored under nitrogen∣The compound is unstable in solutions, freshly prepared is recommended.
Recommended Storage : -20°C, protect from light, stored under nitrogen∣The compound is unstable in solutions, freshly prepared is recommended.
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.