missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilson™ SPE Manifold
SPE Manifold Kit
Brand: Gilson™ 37417013
This item is not returnable.
View return policy
Description
- 15-Position SPE Manifold Kit (6 mL)
- Cartridge rack, SPE, 15-positions, 6 mL (quantity 1)
- Ejection plate for SPE cartridges (quantity 1)
- Gasket with adhesive-backed seal, 15-positions (quantity 2)
- Manifold, SPE, 15-positions (quantity 1)
- O-ring manifold seal (quantity 1)
- Tube rack for SPE collection, 6 mL (quantity 1)
Specifications
15 Position Kit | |
6 mL | |
SPE Manifold |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction