Learn More
J.T. Baker™ Bakerbond spe™ SDB Solid Phase Extraction Columns
Offering excellent structural, mechanical and chemical properties, packaging purity and loadability as well as stability of the SDB sorbent for improved sample clean-up and preconcentration.
Brand: J.T. Baker™ 7519-05
Code : Z3
Additional Details : CAS Number : 9003-53-6 Weight : 0.19000kg
Description
- Highly effective and selective SDB sorbent with high throughput performance
- Enables high recovery and reproducibility of results, even in the SPE of the compounds of different polarities
- 6mL capacity, 200mg bed weight
- 30/pack
Specifications
Styrene-DVB | |
SPE | |
6 mL | |
30/Pk. |