Learn More
3M™ Speaking Diaphragm Kit, 7895
3M™ Speaking Diaphragm Kit, 7895 is our spare speech diaphragm kit designed to replace worn or damaged parts in 3M™ Powerflow™ Full Facepiece 7900.
Brand: 3M™ 7000051802
Code : NEW
Additional Details : Weight : 0.06710kg
Description
Replace old or damaged 3M™ Powerflow™ Full Facepiece 7900 speech diaphragms with 3M™ Speaking Diaphragm Kit, 7895. Our approved 3M replacement part can extend the service life of your facepiece. This kit includes speaking diaphragm, gasket, retainer and removal tool.Specifications
NIOSH Approved | |
Replacement Speaking Diaphragm Kit | |
3M™ Full Facepieces 7000 Series |