missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bochem™ Special 4-Prong Stainless Steel Retort Clamps with Silicone Coating
Use this Bochem™ Special 4-Prong Stainless Steel Retort Clamps with Silicone Coating to handle laboratory glassware as required.
£79.70 - £130.00
Product Code | Brand | Length (Metric) | Diameter (Metric) | Diameter (Metric) Inside | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Diameter (Metric) Inside | Price | Quantity & Availability | |||
10578082
|
Bochem™
5423 |
150 mm | 0 to 120 mm | 0 to 120 mm |
£129.00
Each |
Estimated Shipment: 22-10-2024
Log in to see stock. |
|||
10530052
|
Bochem™
5553 |
190 mm | 20 to 100 mm | 20 to 100 mm |
£79.70
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
12160450
|
Bochem™
5554 |
190 mm | 50 to 150 mm | 50 to 150 mm |
£130.00
Each |
Estimated Shipment: 16-10-2024
Log in to see stock. |
|||
Description
- Type 2
- 18/10 stainless steel
- With silicone coating
Specifications
1 Ea. | |
Stand Clamp | |
Stainless Steel |