Learn More
Invitrogen™ Custom DNA Oligos Synthesis Services
Custom Primer
Brand:  Invitrogen™ 10629012
.15 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
 View more versions of this product
Product Code. 10424182
  • £0.20 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Description
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 nmol
Custom Tube Primer, 50nmole, Purified by Normal or Reversed-phase Chromatography, HPLC or PAGE, Used to Design a Custom DNA Oligonucleotide, With any of a Variety of 3' and 5' Modification
HPLC
Normal or Reversed-phase Chromatography, HPLC or PAGE
Custom Tube Primer
Design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modification