missing translation for 'onlineSavingsMsg'
Learn More

MakerBot™ Specialty Large PLA Filament

MakerBot™ Specialty PLA Filament is composed of the purest grade resin and designed for use with the MakerBot™ 2 Desktop 3D printer and all fifth generation MakerBot™ Replicator 3D printers.

POA - £63.18

Specifications

Product Type PLA Filament
Material PLA
View More Specs

Products 4
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
16301506
Encompass_Preferred
View Documents
MakerBot™
MKS-000031
Glow in the Dark
POA
Each
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

16371496
Encompass_Preferred
View Documents
MakerBot™
MKS-000022
Khaki
POA
Each
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

16381496
Encompass_Preferred
View Documents
MakerBot™
MKS-000023
Army Green
POA
Each
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

16391496
Encompass_Preferred
View Documents
MakerBot™
MKS-000024
Ocean Blue This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Composed of the purest grade resin on the market
  • Comprehensively quality tested
  • Reduces downtime
  • Creates high quality prints
  • Filament Diameter: 1.75 mm
  • Applications: CAD, Conceptual Modeling, Prototyping, Education, AEC
  • Packaging Dimensions (L x W x H): 250 x 250 x 40 mm
Specifications

Specifications

PLA Filament
PLA
Videos
SDS
Documents

Documents

Product Certifications