missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MakerBot™ Specialty Small PLA Filament
MakerBot™ Specialty PLA Filament is composed of the purest grade resin and designed for use with the MakerBot™ 2 Desktop 3D printer and all fifth generation MakerBot™ Replicator 3D printers.
Brand: MakerBot™ MKS-000067
This item is not returnable.
View return policy
Description
- Composed of the purest grade resin on the market
- Comprehensively quality tested
- Reduces downtime
- Creates high quality prints
- Filament Diameter: 1.75 mm
- Applications: CAD, Conceptual Modeling, Prototyping, Education, AEC
- Packaging Dimensions (L x W x H): 148 x 148 x 40 mm
Specifications
Ocean Blue | |
Replicator 2 3D Printer, Replicator+ 3D Printer, Replicator Mini 3D Printer, Replicator Mini+ 3D Printer, Replicator Z18 3D Printer | |
PLA Filament | |
PLA |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction