missing translation for 'onlineSavingsMsg'
Learn More

VEE GEE Specific Gravity Hydrometer, 305 mm

The Precision Specific Gravity Hydrometer is calibrated for use at 60°F (15.55°C).

Brand:  VEE GEE 6603-9

 View more versions of this product

Product Code. 17816623

  • £55.15 / Each
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The specific gravity range and subdivisions differ depending on the model.
Specifications

Specifications

Specific Gravity Hydrometer
30.5 cm
Glass
3.000 to 4.000
0.01
35 x 3 x 3 cm
6 g
60°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
VEE GEE Specific Gravity Hydrometer, 305 mm > Specific Gravity Range: 3.000 to 4.000, Increments: 0.01

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.