missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEE GEE Specific Gravity Hydrometer, 305 mm
The Precision Specific Gravity Hydrometer is calibrated for use at 60°F (15.55°C).
Brand: VEE GEE 6603-2
This item is not returnable.
View return policy
Description
- The specific gravity range and subdivisions differ depending on the model.
Specifications
Specific Gravity Hydrometer | |
30.5 cm | |
Glass | |
1.200 to 1.420 |
0.002 | |
35 x 3 x 3 cm | |
6 g | |
60°F |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VEE GEE Specific Gravity Hydrometer, 305 mm > Specific Gravity Range: 1.200 to 1.420, Increments: 0.02
Spot an opportunity for improvement?Share a Content Correction