missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wheaton™ Specimen Bag with Document Wallet, Biohazard Print
Specimen bag ideal for safe patient sample transport.
Brand: Wheaton™ 370150
91.09 GBP valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 4.57000kg
Description
- Ensures safe sample transfer
- For transportation of clinical specimens and documents
Specifications
Specimen Bag | |
Self-adhesive | |
Transportation of clinical specimens and documents | |
LDPE | |
1000 | |
150 x 150 mm |
Not Autoclavable | |
Single-use | |
Document Pouch, Biohazard Print | |
Non-sterile | |
100 |