missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Specimen Bag with Document Wallet, Biohazard Print

Specimen bag ideal for safe patient sample transport.

Brand:  Wheaton™ 370155

115.21 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 16494979

  • £178.00 / Pack of 1000
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ensures safe sample transfer
  • For transportation of clinical specimens and documents
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Specimen Bag
Self-adhesive
Transportation of clinical specimens and documents
LDPE
1000
150 x 220 mm
Not Autoclavable
Single-use
Document Pouch, Biohazard Print
Non-sterile
50
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.