missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ Specimen Bag with Document Wallet, Plain

Specimen bag ideal for safe patient sample transport.

Brand:  Wheaton™ 370170

110.89 GBP valid until 2024-12-31
Use promo code "23259" to get your promotional price.



Additional Details : Weight : 5.26000kg

 View more versions of this product

Product Code. 16414989

  • £121.00 / Pack of 1000
Estimated Shipment: 22-07-2024
to see stock.
Add to basket

Description

Description

  • Ensures safe sample transfer
  • For transportation of clinical specimens and documents
Specifications

Specifications

Specimen Bag
Self-adhesive
Transportation of clinical specimens and documents
LDPE
1000
150 x 220 mm
Not Autoclavable
Single-use
Document Pouch, Plain
Non-sterile
50
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers