Learn More
Neutral Glass Specimen Tube
Use these tubes for a variety of applications. Neutral Glass Specimen Tubes are ideal for samples which are susceptible to pH change.
Brand: Samco T102/V3/T002/C3
58.29 GBP valid until 2024-12-20
Use promo code "21644" to get your promotional price.
This item is not returnable.
View return policy
Description
- Neutral glass tube with black urea screw cap
- Flat bottom, straight sided
- Good chemical resistance
- Should not be taken from cold and placed directly into the hottest part of the flame without pre-heating
Specifications
28 mm | |
72 mm |