missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum Labs™ Spectra/Mesh™ Woven Stainless Steel Filter Sheet
Hydrophilic stainless steel filter with very high strength for use in laboratory, industrial, or pharmaceutical applications.
Brand: Spectrum Labs™ 148985
Code : NEW
Additional Details : Weight : 0.10000kg
Description
- Hydrophilic
- Good resistance against corrosives
- Resistant against pH 4 to 12
- Sterilize via irradiation/autoclave
Specifications
Stainless Steel | |
Laboratory, Industrial, Pharmaceutical | |
50 μm |
30 μm | |
30% | |
30 x 30 cm |