Learn More
Thermo Scientific™ Spectra™ Multicolor Broad Range Protein Ladder
Thermo Scientific™ Spectra Multicolor Broad Range Protein Ladder is a 4-color protein standard containing 10 prestained proteins (10 to 260 kDa) for use as size standards in gel electrophoresis and Western blotting.
Brand: Thermo Scientific™ 26634
151.65 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : PN
Additional Details : Weight : 1.36080kg
Description
Thermo Scientific Spectra Multicolor Broad Range Protein Ladder is a 4-color (blue, orange, green, pink) protein standard containing 10 prestained proteins (10 to 260 kDa) for use in gel electrophoresis and western blotting. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.
Applications
- Monitoring protein migration during SDS-polyacrylamide gel electrophoresis
- Monitoring protein transfer onto membranes after western blotting
- Sizing of proteins on SDS-polyacrylamide gels and western blots
Specifications
Contents: two vials of 250 μL each |
|
10 | |
Yes | |
4 colors: Blue, Orange, Green, Pink | |
260, 140, 100, 70, 50, 40, 35, 25, 15, 10 kDa | |
2 x 250 μL | |
Western Blotting, SDS-PAGE |
Colorimetric, NIR Fluorescence (700 nm), RGB Fluorescence (555 nm) | |
Protein Ladder | |
10 to 260 kDa | |
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels | |
Spectra™ | |
Approved for shipment on Wet or Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.