missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum Labs™ Spectra/Por™ 1 Membrane Disc
Click to view available options
Diameter (Metric):
47 mm
100 mm
Unit Size:
Pack of 50
Description
For use with Spectrum Equilibrium Dialyzer and Spectra/Por™ MacroDialyzer systems.
Regenerated cellulose (RC) precut membrane discs
Supplied dry with a glycerol as a humectant
Regenerated cellulose (RC) precut membrane discs
Supplied dry with a glycerol as a humectant
Specifications
Specifications
| Product Type | Membrane Disc |
| MWCO | 6 to 8 kDa |
| Membrane | Regenerated Cellulose |
| Diameter (Metric) | 100 mm |
| For Use With (Application) | For Basic Dialysis |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction