Learn More
Spectrum™ Spectra/Por™ Float-A-Lyzer™ G2 Dialysis Devices
Description
Ready-to-use and disposable, Float-A-Lyzer G2 incorporates biotech-grade cellulose ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use and provides ultimate convenience for small volume dialysis (1 to 10mL)
Available in volume sizes: 1mL, 5mL and 10 mL. Resealable screw cap is color-coded for 9 available MWCOs and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5mL and 10mL sizes). Flotation ring (included) provides buoyancy to maximize dialysis efficiency.
- Superior handling and leak protection
- Resealable screw cap and pipet for easy loading and total recovery
- Highest membrane purity, no heavy metals and sulfides
- Minimizes volume specific dilution
- 9 MWCOs ranging from 100 to 1,000,000 Daltons
Specifications
Specifications
| Product Type | Float-A-Lyzer G2 Dialysis Device |
| MWCO | 0.1 to 0.5 kDa |
| Membrane | Cellulose Ester |
| Diameter (English) Membrane | 0.39 in. |
| Diameter (English)Top | 0.9 in. |
| Diameter (Metric) Membrane | 10 mm |
| Diameter (Metric)Top | 23 mm |
| Length (English) | 6.2 in. |
| Length (Metric) | 160 mm |
| Closure Material | Polypropylene |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.