Learn More
Spectrum™ Spectra/Por™ Float-A-Lyzer™ G2 Dialysis Devices
Ready-to-use and disposable, Float-A-Lyzer G2 incorporates biotech-grade cellulose ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use and provides ultimate convenience for small volume dialysis (1 to 10mL)
£152.74 - £314.00
Specifications
Product Type | Float-A-Lyzer G2 Dialysis Device |
---|---|
Membrane | Cellulose Ester |
Diameter (English) Membrane | 0.39 in. |
Diameter (English)Top | 0.9 in. |
Diameter (Metric) Membrane | 10 mm |
Product Code | Brand | Color Code | Length (English) | Length (Metric) | MWCO | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color Code | Length (English) | Length (Metric) | MWCO | Price | Quantity & Availability | ||
11571160
|
Spectrum™
G235035 |
Blue | 1.9 in. | 50 mm | 100 kDa |
£278.00
Pack of 12 |
Estimated Shipment: 08-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11531160
|
Spectrum™
G235029 |
Black | 1.9 in. | 50 mm | 3.5 to 5 kDa |
£202.00
Pack of 12 |
Estimated Shipment: 20-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11541180
|
Spectrum™
G235070 |
Violet | 6.2 in. | 160 mm | 50 kDa |
£189.00
Pack of 12 |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11561170
|
Spectrum™
G235059 |
Blue | 3.9 in. | 100 mm | 100 kDa |
£293.00
Pack of 12 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11581160
|
Spectrum™
G235036 |
Amber | 1.9 in. | 50 mm | 300 kDa |
£283.00
Pack of 12 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11511160
|
Spectrum™
G235025 |
Green | 1.9 in. | 50 mm | 0.1 to 0.5 kDa |
£283.00
Pack of 12 |
Estimated Shipment: 25-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11541160
|
Spectrum™
G235031 |
Yellow | 1.9 in. | 50 mm | 8 to 10 kDa |
£206.00
Pack of 12 |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11551160
|
Spectrum™
G235033 |
Red | 1.9 in. | 50 mm | 20 kDa |
£152.74
Pack of 12 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11581170
|
Spectrum™
G235061 |
Green | 6.2 in. | 160 mm | 0.1 to 0.5 kDa |
£165.00
Pack of 12 |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11591170
|
Spectrum™
G235062 |
Pink | 3.9 in. | 100 mm | 1000 kDa |
£314.00
Pack of 12 |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11561160
|
Spectrum™
G235034 |
Violet | 1.9 in. | 50 mm | 50 kDa |
£278.00
Pack of 12 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11495829
|
Spectrum™
G235067 |
Yellow | 6.2 in. | 160 mm | 8 to 10 kDa |
£189.00
Pack of 12 |
Estimated Shipment: 23-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11571180
|
Spectrum™
G235073 |
Pink | 6.2 in. | 160 mm | 1000 kDa |
£165.00
Pack of 12 |
Estimated Shipment: 05-02-2025 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11501170
|
Spectrum™
G235049 |
Green | 3.9 in. | 100 mm | 0.1 to 0.5 kDa |
£186.00
Pack of 12 |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11501180
|
Spectrum™
G235063 |
Orange | 6.2 in. | 160 mm | 0.5 to 1 kDa |
£232.20
Pack of 12 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11571170
|
Spectrum™
G235060 |
Amber | 3.9 in. | 100 mm | 300 kDa |
£161.00
Pack of 12 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11551170
|
Spectrum™
G235058 |
Violet | 3.9 in. | 100 mm | 50 kDa |
£161.00
Pack of 12 |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11511170
|
Spectrum™
G235051 |
Orange | 3.9 in. | 100 mm | 0.5 to 1 kDa |
£302.38
Pack of 12 |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11591160
|
Spectrum™
G235037 |
Pink | 1.9 in. | 50 mm | 1000 kDa |
£292.06
Pack of 12 |
Estimated Shipment: 23-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11561180
|
Spectrum™
G235072 |
Amber | 6.2 in. | 160 mm | 300 kDa |
£285.00
Pack of 12 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11521160
|
Spectrum™
G235027 |
Orange | 1.9 in. | 50 mm | 0.5 to 1 kDa |
£278.00
Pack of 12 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11541170
|
Spectrum™
G235057 |
Red | 3.9 in. | 100 mm | 20 kDa |
£186.00
Pack of 12 |
Estimated Shipment: 23-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11521170
|
Spectrum™
G235053 |
Black | 3.9 in. | 100 mm | 3.5 to 5 kDa |
£288.00
Pack of 12 |
Estimated Shipment: 01-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11531180
|
Spectrum™
G235069 |
Red | 6.2 in. | 160 mm | 20 kDa |
£170.28
Pack of 12 |
Estimated Shipment: 23-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11837271
|
Spectrum™
G235055 |
Yellow | 3.9 in. | 100 mm | 8 to 10 kDa |
£186.00
Pack of 12 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11827271
|
Spectrum™
G235065 |
Black | 6.2 in. | 160 mm | 3.5 to 5 kDa |
£165.00
Pack of 12 |
Estimated Shipment: 23-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
11551180
|
Spectrum™
G235071 |
Blue | 6.2 in. | 160 mm | 100 kDa |
£290.00
Pack of 12 |
Estimated Shipment: 16-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Ready-to-use and disposable, Float-A-Lyzer G2 incorporates biotech-grade cellulose ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use and provides ultimate convenience for small volume dialysis (1 to 10mL)
Available in volume sizes: 1mL, 5mL and 10 mL. Resealable screw cap is color-coded for 9 available MWCOs and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5mL and 10mL sizes). Flotation ring (included) provides buoyancy to maximize dialysis efficiency.
- Superior handling and leak protection
- Resealable screw cap and pipet for easy loading and total recovery
- Highest membrane purity, no heavy metals and sulfides
- Minimizes volume specific dilution
- 9 MWCOs ranging from 100 to 1,000,000 Daltons
Specifications
Float-A-Lyzer G2 Dialysis Device | |
0.39 in. | |
10 mm | |
Polypropylene | |
Disposable pipette for 5 and 10mL, flotation ring | |
Ready to use laboratory dialysis devices, highest membrane purity, superior handling and leak prevention, 95 to 98% sample recovery, perfect for dialyzing electro-eluted proteins or chromatography fractions |
Cellulose Ester | |
0.9 in. | |
23 mm | |
Screw Cap | |
12 Pack |