Learn More
Spectrum Labs™ SpectraPor™ Tube-A-Lyzer™ Dialysis Device
The SpectraPor™ Tube-A-Lyzer™ Dialysis Device is a self-contained, ready to use separation device that combines convenience and disposability, the dynamic dialysis increases the mass transfer rate and reduces overall dialysis from 1 to 2 days to 12 hours.
Brand:  Spectrum Labs™ 137050
 View more versions of this product
Product Code. 11521110
  • £256.00 / Pack of 3
Explore more special offers
Description
Description
A revolutionary disposable dialysis device designed for the rapid yet gentle dialysis and separation of larger sample volumes; exhibiting higher efficiency as well as product recovery. Available in 6 MWCOs (0.1 to 0.5 kD, 3.5 to 5 kD, 8 to 10 kD, 20 kD, 50 kD and 100 kD) and two sample volume sizes (8 to 10 mL and 25 to 28 mL).
Tube-A-Lyzer™ is a self-contained ready to use mass transfer system that incorporates a semi-permeable, tubular dialysis membrane inside a clear reservoir housing to separate the sample chamber from the surrounding buffer chamber. The high purity Biotech Cellulose Ester (CE) membrane is dry, impregnated with 20% glycerine, and offers high molecular selectivity. The full length of the membrane tubing has a working volume of approximately 28 mL (Special length units for larger or smaller volumes can be made on custom build basis). A female Luer Lock port is located on the top for sample loading, recovery or in-process testing from the membrane enclosed sample chamber. Designed for dynamic dialysis, the housing (buffer) chamber is equipped with 6 mm inlet and outlet hose-barb ports that can be connected to flexible plastic tubing (3/16 x 5/16 inch I.D.) for recirculation of a larger buffer volume using a peristaltic pump. Alternatively, the ports can be capped off to facilitate only the buffer chamber for static (standard) dialysis. Multiple Tube-A-Lyzer™ devices can be connected in parallel to create a larger sample volume manifold.
  • Maintains maximum concentration gradient
  • Prevents polarisation and membrane fouling
  • Provides hypo-osmic pressure to aid separation
  • Achieves higher sample purities
  • Reduces time from days to hours
  • Ready to use and disposable
  • Sterile and non-pyrogenic
  • Biotech Grade RC and CE membrane (0.1kD to 100kD)
  • Large volume sample chamber
  • Recirculating dialysate for rapid dialysis
Typical applications:
Desalting; Concentration of plasma/serum; Buffer and pH change; Protein preparation for electrophoresis; Concentration of antibodies; Removal of contaminant; Binding studies; Flow dialysis/batch analysis; Temperature regulated dialysis; Tissue culture extract purification; Protein elution for electrophoresis gel; Remove oligosaccharides from protein solution.
Specifications
Specifications
Tubular Dialysis Membrane
100 kDa
Biotech Cellulose Ester
25 to 30 mL