Learn More
Thermo Scientific™ SPECTRONIC 200™ Spectrophotometer
Thermo Scientific™ SPECTRONIC™ 200 is the new standard for rugged, simple-to-use visible spectrophotometers for teaching and routine applications.
£2265.00 - £2430.00
Specifications
Compartments Removable, washable sample compartment
Connections USB-B for connection to a remote computer
USB-A for connection to a printer or USB memory device
Container Type Single cell holder for square cuvettes and for test tubes
Data Interval for Scanning 1 nm on-board, 0.5 nm under computer control
Depth (English) 11.8 in.
View More Specs
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15589673
View Documents
Thermo Scientific™
840-281800
SPECTRONIC 200 spectrophotometer with UK Power Cord
£2265.00
Each
15591353
View accessory products View Documents
Thermo Scientific™
840-281900
SPECTRONIC 200 spectrophotometer with Euro Power Cord
£2430.00
Each
Description
Show More
Specifications
Show More
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ SPECTRONIC 200™ Spectrophotometer