missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reagecon™ Spectrophotometric Standards
Calibrate a spectrophotometer to ensure accurate transmission measurements. Spectrophotometric Standards, Reagecon™ are safe, stable and easy-to-use optical standards with guaranteed accuracy of ±5%. All solutions are NIST traceable and come with a certificate of analysis.
Brand: Reagecon™ OPS30
This item is not returnable.
View return policy
Description
Spectrophotometry is the quantitative measurement of the reflection or transmission properties of a material. Spectrophotometers are used to measure a light beam's intensity as a function of its color (wavelength). These reference standards act as a calibration and control solution, with highly accurate absorbance data at a variety of wavelengths.
- NIST Traceable—Instruments used to acquire data are NIST calibrated
- Easy to Use—Pour straight out of the plastic bottle
- Accurate—Guaranteed accuracy of ±5% inter/intra-instrument variance
- Stable—Will not settle out of solution, guaranteed one year shelf life
- Safe—Non carcinogenic materials used
Specifications
0.1929 to 2.6225 A | |
Optical standard 30.0 | |
Calibration of spectrophotometers | |
125 mL Plastic Bottles |
NIST traceable | |
450 to 224 nm | |
Certificate of analysis | |
Shelf life: 1 year |