Learn More
Reagecon™ Spectrophotometric Standards
Calibrate a spectrophotometer to ensure accurate transmission measurements. Spectrophotometric Standards, Reagecon™ are safe, stable and easy-to-use optical standards with guaranteed accuracy of ±5%. All solutions are NIST traceable and come with a certificate of analysis.
£277.00 - £391.00
Specifications
Certifications/Compliance | NIST traceable |
---|---|
Wavelength Range | 450 to 224 nm |
For Use With (Application) | Calibration of spectrophotometers |
Includes | Certificate of analysis |
Packaging | 125 mL Plastic Bottles |
Product Code | Brand | Absorbance Range | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Absorbance Range | Description | Price | Quantity & Availability | ||||
11744815
|
Reagecon™
OPS1 |
0.0058 to 0.094 A | Optical standard 1.0 | ||||||
11754815
|
Reagecon™
OPS2 |
0.012 to 0.02023 A | Optical standard 2.0 | ||||||
11794815
|
Reagecon™
OPS30 |
0.1929 to 2.6225 A | Optical standard 30.0 | ||||||
11774815
|
Reagecon™
OPS8 |
0.049 to 0.7928 A | Optical standard 8.0 | ||||||
Description
Spectrophotometry is the quantitative measurement of the reflection or transmission properties of a material. Spectrophotometers are used to measure a light beam's intensity as a function of its color (wavelength). These reference standards act as a calibration and control solution, with highly accurate absorbance data at a variety of wavelengths.
- NIST Traceable—Instruments used to acquire data are NIST calibrated
- Easy to Use—Pour straight out of the plastic bottle
- Accurate—Guaranteed accuracy of ±5% inter/intra-instrument variance
- Stable—Will not settle out of solution, guaranteed one year shelf life
- Safe—Non carcinogenic materials used
Specifications
NIST traceable | |
Calibration of spectrophotometers | |
125 mL Plastic Bottles |
450 to 224 nm | |
Certificate of analysis | |
Shelf life: 1 year |